Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Millistak+™ HC Pod Disposable Depth Filter System

Catalog No. MA1HC027H1 Shop All MilliporeSigma Products
Change view
Click to view available options
Grade:
A1HC
B1HC
C0HC
D0HC
Filtration Area:
0.027 m2
0.054 m2
For Use With (Application):
Perfusion bioreactor fluid, laboratory filtration
Post-TFF clarification fluids, laboratory filtration
Post-centrifuge or settled permeate containing cellular particulate, laboratory filtration
Primary clarification directly out of the bioreactor, laboratory filtration
7 product options available for selection
Product selection table with 7 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Grade Filtration Area For Use With (Application)
MA1HC027H1 A1HC 0.027 m2 Post-TFF clarification fluids, laboratory filtration
MA1HC054H1 A1HC 0.054 m2 Post-TFF clarification fluids, laboratory filtration
MB1HC027H1 B1HC 0.027 m2 Post-centrifuge or settled permeate containing cellular particulate, laboratory filtration
MB1HC054H1 B1HC 0.054 m2 Post-centrifuge or settled permeate containing cellular particulate, laboratory filtration
MC0HC027H1 C0HC 0.027 m2 Perfusion bioreactor fluid, laboratory filtration
MD0HC054H1 D0HC 0.054 m2 Primary clarification directly out of the bioreactor, laboratory filtration
MD0HC027H1 D0HC 0.027 m2 Primary clarification directly out of the bioreactor, laboratory filtration
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 7 options available.
7 options
Catalog No. MA1HC027H1 Supplier MilliporeSigma™ Supplier No. MA1HC027H1
Add to Cart
Add to Cart

Combine two distinct technologies to enhance filter capacity and retention

  • Filter material: cellulose and inorganic filter aid
  • Configurations: 0.11, 0.55, or 1.1 m2 filtration area
  • Low hold-up volume for greater product yield
  • Broad range of 23 media types offered in single and multilayer products
  • Flexible, modular format offers scalability from 5 to 12,000L or more
  • Patented disposable design eliminates need for housing, CIP or cleaning validation
  • Disposable Pod system protects operators from exposure to biohazards
  • Robust construction is easy to use and set up
  • Smaller footprint facilitates use in tight spaces

Ideal for Primary and Secondary Clarification in Lab, Pilot and Process-Scale Applications

TRUSTED_SUSTAINABILITY

Specifications

Disposable Single-use
Material Glass Fiber-reinforced Polypropylene
Material (Membrane) Cellulose
Filtration Area 0.027 m2
For Use With (Application) Post-TFF clarification fluids, laboratory filtration
Grade A1HC
Inlet Connection (English) 0.25 in. hose
Max. Pressure 50 psi at 25°C, 15 psi at 80°C
No. per Pack 1 Ea.
Type Millistak+ Lab Scale Pod Filter
Length (English) 8.5 in.
Length (Metric) 22 cm
Height (English) 5.3 in.
Height (Metric) 14 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.