Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More
Learn More
- Home
- Products
- Filters and Filtration
- Capsules and Inline Filters
- MilliporeSigma™ Millistak+™ Pod Disposable Depth Filter System
MilliporeSigma™ Millistak+™ Pod Disposable Depth Filter System
Catalog No. MCE40054H1 Shop All MilliporeSigma Products
Change view
Click to view available options
Grade:
CE15
CE20
CE25
CE30
CE35
CE40
CE45
CE50
DE25
DE30
DE35
DE40
DE45
DE50
DE55
DE60
DE65
DE70
DE75
Filtration Area:
0.027 m2
0.054 m2
Material (Membrane):
Cellulose
Cellulose, Inorganic Filter Aid
38 product options available for selection
| Catalog No. | Grade | Filtration Area | Material (Membrane) |
|---|---|---|---|
| MCE40054H1 | CE40 | 0.054 m2 | Cellulose |
| MCE15054H1 | CE15 | 0.054 m2 | Cellulose |
| MCE20027H1 | CE20 | 0.027 m2 | Cellulose |
| MCE15027H1 | CE15 | 0.027 m2 | Cellulose |
| MCE20054H1 | CE20 | 0.054 m2 | Cellulose |
| MCE25027H1 | CE25 | 0.027 m2 | Cellulose |
| MCE25054H1 | CE25 | 0.054 m2 | Cellulose |
| MCE30054H1 | CE30 | 0.054 m2 | Cellulose |
| MCE35054H1 | CE35 | 0.054 m2 | Cellulose |
| MCE30027H1 | CE30 | 0.027 m2 | Cellulose |
| MCE40027H1 | CE40 | 0.027 m2 | Cellulose |
| MCE35027H1 | CE35 | 0.027 m2 | Cellulose |
| MCE50027H1 | CE50 | 0.027 m2 | Cellulose |
| MDE25027H1 | DE25 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
| MDE25054H1 | DE25 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
| MDE30054H1 | DE30 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
| MDE35054H1 | DE35 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
| MDE35027H1 | DE35 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
| MDE30027H1 | DE30 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
| MDE40054H1 | DE40 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
| MDE40027H1 | DE40 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
| MDE45027H1 | DE45 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
| MDE50027H1 | DE50 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
| MDE45054H1 | DE45 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
| MDE50054H1 | DE50 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
| MDE60027H1 | DE60 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
| MDE55054H1 | DE55 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
| MDE55027H1 | DE55 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
| MDE65027H1 | DE65 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
| MDE60054H1 | DE60 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
| MDE75054H1 | DE75 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
| MDE70054H1 | DE70 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
| MDE65054H1 | DE65 | 0.054 m2 | Cellulose, Inorganic Filter Aid |
| MDE70027H1 | DE70 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
| MDE75027H1 | DE75 | 0.027 m2 | Cellulose, Inorganic Filter Aid |
| MCE45027H1 | CE45 | 0.027 m2 | Cellulose |
| MCE50054H1 | CE50 | 0.054 m2 | Cellulose |
| MCE45054H1 | CE45 | 0.054 m2 | Cellulose |
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 38 options available.
Description
- Filter Material: cellulose
- Configurations: 0.11, 0.77, or 1.4 m2 filtration area
- Low hold-up volume for greater product yield
- Broad range of 23 media types offered in single and multilayer products
- Flexible, modular format offers scalability from 5 to 12,000L or more
- Patented disposable design eliminates need for housing, CIP or cleaning validation
- Disposable Pod system protects operators from exposure to biohazards
- Robust construction is easy to use and set up
- Smaller footprint facilitates use in tight spaces
Specifications
Specifications
| Disposable | Single-use |
| Material | Glass Fiber-reinforced Polypropylene |
| Material (Membrane) | Cellulose |
| Filtration Area | 0.054 m2 |
| For Use With (Application) | Clarification of serum, plasma, vaccines, cell culture or other fluids |
| Grade | CE40 |
| Inlet Connection (English) | 0.5 in. hose barb |
| Max. Pressure | 50 psi at 25°C, 15 psi at 80°C |
| No. per Pack | 1 Ea. |
| Type | Millistak+ Pod Depth Filter |
| Volume (Metric) | 5 to 12,000 L |
| Height (English) | 5.3 in. |
| Height (Metric) | 13.46 cm |
| Show More Show Less |
Product Title
Select an issue
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?