Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MKK6, GST-Fusion, Human, Recombinant Protein, E. coli

Catalog No. 47570120UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
20 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-570-120UG 20 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-570-120UG Supplier MilliporeSigma™ Supplier No. 47570120UG
Recombinant, human MKK6 fused to GST at the N-terminus to GST and expressed in E. coli. MKK6 is a member of the dual specificity protein kinase family, which functions as a mitogen-activated protein (MAP) kinase kinase. It is suitable for labeling MAP kinase substrates.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 270mM sucrose, 150mM NaCl, 50mM Tris-HCl, 1mM DTT, 0.1mM EGTA, pH 7.5.
Purification Method SDS-PAGE
Quantity 20 μg
Source E. coli
Immunogen MKK6
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Conjugate GST-Fusion
Species Human
Recombinant Recombinant
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.