Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MMP-2, Proenzyme, Human, Recombinant Protein, CHO Cells

Catalog No. PF03710UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PF03710UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PF03710UG Supplier MilliporeSigma™ Supplier No. PF03710UG
Add to Cart
Add to Cart
Recombinant, human pro-MMP-2 expressed in CHO cells. Can be used as a positive control for zymographic analysis, immunoblotting, and substrate cleavage assay. A portion (less than 10%) of the enzyme may be seen as a naturally-occurring dimer. Dimerization does not interfere with activation. May contain up to 10% TIMP proteins. During storage, a small portion (less than 10%) of the enzyme may also become activated.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 150mM NaCl, 20mM Tris-HCl, 5mM CaCl2, 0.05% BRIJ™-35 Detergent, pH 7.4.
Purification Method SDS-PAGE
Quantity 10 μg
Immunogen MMP-2
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Cell Type CHO Cells
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.