Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MMP-3, Proenzyme, Human, Recombinant Protein

Catalog No. PF06310UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
PF06310UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. PF06310UG Supplier MilliporeSigma™ Supplier No. PF06310UG
Add to Cart
Add to Cart
Recombinant, human pro-MMP-3 purified from cell culture supernatant. May be used as a positive control or standard for zymographic analysis, or substrate assay. Requires activation for immunoblotting, prior to use. M.W. 57000/58000.
TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) Immunoblotting, Substrate Cleavage Assay, Zymography
Formulation Lyophilized from 100mM NaCl, 50mM HEPES, pH 7.3.
Preparation Method H2O
Purification Method SDS-PAGE
Quantity 10 μg
Immunogen MMP-3
Storage Requirements Following reconstitution, aliquot into siliconized vials and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.