Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Monocyte Chemotactic Protein-1, Human, Recombinant Protein, E. coli

Catalog No. 47590110UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-590-110UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-590-110UG Supplier MilliporeSigma™ Supplier No. 47590110UG
Recombinant, human monocyte chemotactic protein-1 expressed in E. coli. A potent chemoattractant for human monocytes and mouse peritoneal macrophages. Its effects are proposed to be mediated by P2-integrins.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from sterile-filtered PBS, 50μg BSA/μg MCP-1.
Preparation Method Reconstitute to a concentration ≥10μg/mL with sterile PBS containing at least 0.1% HSA or BSA.
Purification Method SDS-PAGE
Quantity 10 μg
Source E. coli
Immunogen Monocyte Chemotactic Protein-1
Storage Requirements Following reconstitution, aliquot and freeze (−70°C) for long term storage or refrigerate (4°C) for short term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 1 year at −70°C.
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.