Learn More
Description
Specifications
Specifications
| Applications | ELISA |
| Concentration | 2.0 mg/mL |
| Description | 2.0 mg/mL |
| Formulation | Purified immunoglobulin. Liquid (sterile filtered), in 0.1M Tris Chloride, 0.5M NaCl pH 8.0 with 0.1% sodium azide. |
| Quantity | 1 mg |
| Content And Storage | 2°C to 8°C as an undiluted liquid for up to 12 months. For extended storage mix with an equal volume of glycerol (ACS grade or better), aliquot contents and freeze at -20°C or below for up to 6 months. Avoid repeated freeze/thaw cycles. |
| Product Type | IgM |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.