Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MQuant™ Nitrate Test Strips
GSA_VA

Catalog No. M1100200002 Shop All MilliporeSigma Products
Change view
Click to view available options
No. of Tests:
25
100
1000
3 product options available for selection
Product selection table with 3 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. of Tests
M1100200002 25
M1100200001 100
M1100920021 1000
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 3 options available.
3 options
Catalog No. M1100200002 Supplier MilliporeSigma™ Supplier No. 1.10020.0002
Add to Cart
Add to Cart

Includes

Tube containing 100 strips

To determine nitrate in aqueous media and food

  • Small and easy to handle
  • Flexible and universal application
  • Modified Griess' reaction method ideal for screening
  • Immediate result produced through on-site analysis
  • PET film and low reagent content
  • Easily disposable
  • Color-scale graduation: 10 - 25 - 50 - 100 - 250 - 500mg/L

Reclose the tube containing the test strips immediately after use.

For food and water analysis

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) For food and water analysis
For Use With (Equipment) Colorimeter
Graduations 10-25-50-100-250-500mg/L
No. of Tests 25
Range 10 to 500mg/L
Storage Requirements 2°C to 8°C
Test Method Modified Griess' reaction
Type Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.