Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MQuant™ Nitrite Test Strips

Catalog No. M1100220001 Shop All MilliporeSigma Products
Change view
Click to view available options
Range:
0.1 to 3g/L
0.5 to 10mg/L NO2-
2 to 80mg/L
2 to 80mg/L NO2-
Graduations:
0.1-0.3-0.6-1-2-3g/L
0.5-1-2-5-10mg/L
2-5-10-20-40-80mg/L
No. of Tests:
25
75
100
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Range Graduations No. of Tests
M1100220001 0.1 to 3g/L 0.1-0.3-0.6-1-2-3g/L 100
M1100070001 2 to 80mg/L 2-5-10-20-40-80mg/L 100
M1100070002 2 to 80mg/L NO2- 2-5-10-20-40-80mg/L 25
M1100570001 0.5 to 10mg/L NO2- 0.5-1-2-5-10mg/L 75
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. M1100220001 Supplier MilliporeSigma™ Supplier No. 1.10022.0001

Includes

Tube containing 100 strips

To determine nitrite in analysis of water and cooling lubricants

  • Small and easy to handle
  • Flexible and universal application
  • Griess' reaction method ideal for screening
  • Immediate result produced through on-site analysis
  • PET film and low reagent content
  • Easily disposable

1.10007.0001
Color-scale graduation: 2 - 5 - 10 - 20 - 40 - 80mg/L

1.10022.0001
Color-scale graduation: 0.1 - 0.3 - 0.6 - 1 - 2 - 3g/L

Reclose the tube containing the test strips immediately after use.

For food and water analysis

TRUSTED_SUSTAINABILITY

Specifications

For Use With (Application) To determine nitrite in analysis of water and cooling lubricants
For Use With (Equipment) Colorimeter
Graduations 0.1-0.3-0.6-1-2-3g/L
No. of Tests 100
Range 0.1 to 3g/L
Storage Requirements 2°C to 8°C
Test Method Griess' reaction
Type Test Strip
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.