Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MT1-MMP, Catalytic Domain, Human, Recombinant Protein, E. coli

Catalog No. 47593510UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
47-593-510UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 47-593-510UG Supplier MilliporeSigma™ Supplier No. 47593510UG
Add to Cart
Add to Cart
Recombinant, human pro-MT1-MMP expressed in E. coli and activated to yield the active catalytic domain (amino acids 89-265). Degrades fibrillar collagen Types I, II, and III, as well as other extracellular matrix proteins, and activates MMP-2 and MMP-13 resulting in enhanced proteolysis of the matrix for cell migration and proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 150mM NaCl, 50mM Tris-HCl, 5mM CaCl2, pH 7.5.
Purification Method SDS-PAGE
Quantity 10 μg
Source E. coli
Immunogen MT1-MMP
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.