Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreen™ HTS 96-Well Filter Plates with MCE Membrane
GSA_VA

Catalog No. MSHAS4510 Shop All MilliporeSigma Products
Change view
Click to view available options
Color:
Clear
Opaque
Material:
Barex
Styrene
Sterility:
Non-sterile
Sterile
Quantity:
10/Pk.
50/Pk.
5 product options available for selection
Product selection table with 5 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Color Material Sterility Quantity
MSHAS4510 Clear Styrene Sterile 10/Pk.
MSHAN4510 Clear Styrene Non-sterile 10/Pk.
MSHAN4550 Clear Styrene Non-sterile 50/Pk.
MSHAN4B50 Opaque Barex Non-sterile 50/Pk.
MSHAS4B10 Opaque Barex Sterile 10/Pk.
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 5 options available.
5 options
Catalog No. MSHAS4510 Supplier MilliporeSigma™ Supplier No. MSHAS4510
Add to Cart
Add to Cart

Commonly used for Elispot assays, bacterial screening, ELISA or viral infection studies

  • Developed for high-throughput use with automated work stations
  • Individually sealed wells prevent incubation crosstalk
  • Removable underdrain allows access to the filters
  • Rigid sidewalls improve handling
  • Ample surfaces provided for bar code labels
  • Pore size: .45μm

Solid-phase RIA, DNA hybridization, ELISA and protein binding

This product(s) resides on a Fisher Scientific GSA or VA contract. If you are viewing this page as a nonregistered user, the price(s) displayed is List Price. To view your GSA or VA contract pricing, log in using your account number, or become a registered user by contacting one of our Customer Service teams. You can also view your contract price by searching for this item(s) on GSA Advantage. To place an order, contact Fisher Scientific Customer Service.

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Membrane Mixed Cellulose Esters
Array 8 x 12
Pore Size 0.45 μm
Color Clear
Material Styrene
No. of Wells 96
Sterility Sterile
Filtration Area 0.28 cm2
For Use With (Application) Solid phase RIA, DNA hybridization, ELISA, protein binding
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.