Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreen™-Mesh Filter Plates with 96-Well Receiver Plates

Catalog No. MANMN2010 Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size:
20 μm
40 μm
60 μm
100 μm
4 product options available for selection
Product selection table with 4 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
MANMN2010 20 μm
MANM10010 100 μm
MANMN4010 40 μm
MANMN6010 60 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 4 options available.
4 options
Catalog No. MANMN2010 Supplier MilliporeSigma™ Supplier No. MANMN2010
Add to Cart
Add to Cart

Ideal for target screening and other applications that evaluate new compounds using multicellular organisms as the in vivo model

96-well plate and 96-well receiver tray fit together to provide a complete system for culturing, testing, and reading assay results in a one-step protocol. Simply transfer parasites, nematodes or other multicellular organisms into the wells, treat with target compound, and measure the migration of the organisms.

  • Provide a ready-to-use system for assays measuring paralysis, cytotoxicity, and death of multicellular organisms
  • Optically clear for use with an imaging microscope or other microscopic analysis
  • Can be coated with a thin layer of agar if necessary
  • Have also been shown to be useful as high-throughput cell strainers prior to cellular analysis in a FACS™ analyzer

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Membrane Nylon Mesh
Array 8 x 12
Pore Size 20 μm
Color Clear
Material Styrene
No. of Wells 96
Sterility Non-sterile
Filtration Area 0.28 cm2
For Use With (Application) Whole organism screening
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.