Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreen™ Solvinert Filter Plates

Catalog No. MDRPN0410 Shop All MilliporeSigma Products
Change view
Click to view available options
Membrane:
Hydrophilic PTFE
Hydrophobic PTFE
Height (Metric):
1.44 cm
1.46 cm
4.07 cm
No. per Pack:
10/Pk.
50
6 product options available for selection
Product selection table with 6 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Membrane Height (Metric) No. per Pack
MDRPN0410 Hydrophobic PTFE 4.07 cm 10/Pk.
MDRLN0410 Hydrophilic PTFE 4.07 cm 10/Pk.
MSRLN0410 Hydrophilic PTFE 1.46 cm 10/Pk.
MSRLN0450 Hydrophilic PTFE 1.46 cm 50
MSRPN0410 Hydrophobic PTFE 1.44 cm 10/Pk.
MSRPN0450 Hydrophobic PTFE 1.46 cm 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 6 options available.
6 options
Catalog No. MDRPN0410 Supplier MilliporeSigma™ Supplier No. MDRPN0410
Add to Cart
Add to Cart

Optimized for drug discovery applications. MultiScreen™ Solvinert Filter Plates are solvent compatible and feature a unique one-piece design. 96-well plates are available in deep well (2mL) format (with or without pre-filter) and standard (500μL) format, with hydrophobic or hydrophilic membranes.

  • Clear polyolefin base plate offers broad solvent compatibility
  • One-piece design eliminates leaking and allows for extended incubations
  • Low binding and low extractables
  • Accommodate aqueous and nonaqueous samples with chemically resistant hydrophilic or hydrophobic membranes
  • Polytetrafluoroethylene membranes feature 45μL pores
  • Automation compatible
  • Available in 96-well deep well (2mL) format (with or without pre-filter) and standard (500μL) format
TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Membrane Hydrophobic PTFE
Array 8 x 12
Pore Size 0.45 μm
Color Clear
Material Polyolefin Copolymer
No. of Wells 96
Sterility Non-sterile
Filtration Area 0.28cm2
For Use With (Application) Solid phase synthesis and cleanup, in plate protein precipitation, sample recovery following in-plate compound cleavage, peptide synthesis
No. per Pack 10/Pk.
Quantity 10/Pk.
Length (Metric) 1.28 cm
Width (Metric) 8.55 cm
Height (Metric) 4.07 cm
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.