Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreenHTS 96-Well Filter Plates

Catalog No. MSIPS4510 Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size:
0.45 μm
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size
MSIPS4510 0.45 μm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Catalog No. MSIPS4510 Supplier MilliporeSigma™ Supplier No. MSIPS4510
Add to Cart
Edge
Add to Cart

missing translation for 'validMainframeMsg'

This item is not returnable. View return policy

For filtration-based solubility determination. MultiScreenHTS 96-Well Filter Plates are specifically developed for use with automation.

  • High Throughput
  • Meet ANSI/SBS 2004 plate specifications
  • Protects underdrain, eliminates surface contact with individual well ‘drip directors’
  • Plastic skirt enables stacking directly with collection plates

Applications

  • Receptor-ligand binding; general sample preparation; Elispot; enzyme activity; bead-based assays; whole organism visualisation
  • Radioisotope detection
TRUSTED_SUSTAINABILITY

Specifications

Certifications/Compliance ANSI, SBS
Membrane Hydrophobic PVDF
Array 8 x 12
Pore Size 0.45 μm
Color Clear
Material ABS Plastic
No. of Wells 96
Sterility Sterile
Filtration Area 0.28cm2
For Use With (Application) ELISPOT assays
Quantity 10/Pk.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.