Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ MultiScreen™HTS 96-Well Filter Plates for Clearing

Catalog No. MSNANLY10 Shop All MilliporeSigma Products
Change view
Click to view available options
No. per Pack:
10
50
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. No. per Pack
MSNANLY10 10
MSNANLY50 50
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. MSNANLY10 Supplier MilliporeSigma™ Supplier No. MSNANLY10
Add to Cart
Add to Cart

Designed to meet specific performance criteria for sample preparation methodology

  • Automation-compatible multiscreen filter plates
  • Lysate clearing

Clearing of bacterial cell lysates

TRUSTED_SUSTAINABILITY

Specifications

Format Standard
Array 8 x 12
Color Clear
Material Styrene
No. of Wells 96
Sterility Non-sterile
For Use With (Application) Clearing of bacterial cell lysates
No. per Pack 10
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.