Learn More
Description
- Automation-compatible multiscreen filter plates
- Lysate clearing
Clearing of bacterial cell lysates
Specifications
Specifications
| Format | Standard |
| Membrane | Negatively Charged Phosphocellulose, Polyester Mesh |
| Array | 8 x 12 |
| Pore Size | 0.65 μm |
| Color | Clear |
| Material | Barex |
| No. of Wells | 96 |
| Sterility | Non-sterile |
| Filtration Area | 0.26 cm2 |
| For Use With (Application) | Protein kinase assays with peptide substrates, enzyme assays, binds positive charge |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.