Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ N-Glycosidase F, Elizabethkingia meningosepticum, Protein

Catalog No. 362185100U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
36-218-5100U 100U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 36-218-5100U Supplier MilliporeSigma™ Supplier No. 362185100U
Add to Cart
Add to Cart
Native N-Glycosidase F from Elizabethkingia meningosepticum. Catalyzes the hydrolysis of asparagine-linked high mannose, as well as hybrid and complex oligosaccharides from glycoproteins. This enzyme will not cleave oligosaccharides containing α1,3-linked core fucose commonly found on plant glycoproteins. N-Glycosidase F will also fail to cleave if the asparagine to which the oligosaccharide is attached is not peptide-bonded to at least one amino acid residue at both the amino and carboxy termini.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 50mM NaCl, 20mM Tris-HCl, 1mM EDTA, pH 7.5.
Quantity 100U
Source Elizabethkingia meningosepticum
Immunogen N-Glycosidase F
Regulatory Status RUO
Recombinant Native
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.