Learn More
Description
Specifications
Specifications
| Preparation Method | Reconstitute to 5 units/mL in 30mM potassium phosphate buffer, pH 7.4. Dilute in 30mM potassium phosphate buffer, pH 7.4, with 0.1% BSA just prior to use. |
| Quantity | 5 U |
| Source | Bacillus licheniformis |
| Immunogen | NADH Oxidase |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C) for long-term storage or store at room temperature (20°C) for short-term storage. Stock solutions are stable for up to 1 week at 20°C or for up to 6 months at −20°C. |
| Regulatory Status | RUO |
| Recombinant | Native |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.