Learn More
Specifications
Specifications
| Concentration | 0.5mg/mL |
| Formulation | Purified NCAM in 15mM octylthioglucoside/ 0.5M NaHCO3. Contains no preservative. |
| Gene ID (Entrez) | XM_001234121.1 |
| Name | Neural Cell Adhesion Molecule (NCAM1), chicken |
| Quantity | 100 μg |
| Storage Requirements | -20°C in undiluted aliquots for up to 6 months. Avoid repeated freeze/thaw cycles. If required for in vivo applications, detergent can be dialyzed away. |
| Form | Purified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.