Learn More
Specifications
Specifications
| Concentration | 100μg/mL |
| For Use With (Application) | Inhibition of cell-cell adhesion, Cell-ECM Adhesion and Cell Migration, ELISA |
| Formulation | Purified Neurocan in PBS. Contains no preservative. |
| Gene ID (Entrez) | NM_004386.2 |
| Name | Neurocan, chicken |
| Quantity | 10 μg |
| Storage Requirements | -20°C in undiluted aliquots for up to 12 months. Avoid repeated freeze/thaw cycles. |
| Form | Purified |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.