Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Nitric Oxide Synthase, Inducible, Mouse, Recombinant Protein, E. coli

Catalog No. 48277750U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
48-277-750U 50 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 48-277-750U Supplier MilliporeSigma™ Supplier No. 48277750U
Add to Cart
Add to Cart
Recombinant, mouse inducible NOS expression in E. coli.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 50mM HEPES, 10% glycerol, pH 7.4.
Quantity 50 U
Source E. coli
Immunogen Nitric Oxide Synthase
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Mouse
Recombinant Recombinant
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.