Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ 10mM dNTP Mix

Catalog No. 710043MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.2mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-004-3MI 0.2mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-004-3MI Supplier MilliporeSigma™ Supplier No. 710043
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Qualified for enzymatic DNA synthesis

The 10mm dNTP Mix is a ready-to-use preparation of ultrapure dATP, dCTP, dGTP, and dTTP (monosodium salts) at a concentration of 10mm each in sterile deionized water at pH 7.0. The dNTP Mix is free of RNase and DNase and is qualified for any application that requires pure deoxynucleotides, such as PCR, cDNA synthesis, and fill-in reactions.
TRUSTED_SUSTAINABILITY

Specifications

Concentration 10 mM
Content And Storage -20°C
Product Type dNTP Mix
Quantity 0.2mL
For Use With (Application) Standard PCR, cDNA Synthesis
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.