Learn More
MilliporeSigma™ Novagen™ NovaBlue T1R Singles™ Competent Cells
Catalog No. 713184 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
22 Reactions
Catalog No. Quantity
71-318-4 22 Reactions
1 options
Catalog No. 71-318-4 Supplier MilliporeSigma™ Supplier No. 713184
Add to Cart
Add to Cart
Featuring T1 and T5 phage resistance
NovaBlue T1R Singles™ Competent Cells have all the same features of NovaBlue Singles, with the added benefit of being resistant to T1 and T5 phage.
TRUSTED_SUSTAINABILITY
Specifications
Efficiency Guaranteed efficiency: >1.5 x 108 cfu/μg
Content And Storage ≤-70°C
Quantity 22 Reactions