Learn More
MilliporeSigma™ Novagen™ NovaTaq™ PCR Master Mix
Catalog No. 710073MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
200 Reactions
Catalog No. Quantity
71-007-3MI 200 Reactions
1 options
Catalog No. 71-007-3MI Supplier MilliporeSigma™ Supplier No. 710073
Add to Cart
Add to Cart
2X PCR reaction components for convenience and reproducibility
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage 2X mixture of NovaTaq DNA Polymerase, ultrapure deoxynucleotides, reaction buffer without MgCl2. Store at -20°C
Quantity 200 Reactions