Learn More
MilliporeSigma™ Novagen™ Origami™ B Competent Cell Set
Catalog No. 709113 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
Catalog No. Quantity
70-911-3 1 Set
1 options
Catalog No. 70-911-3 Supplier MilliporeSigma™ Supplier No. 709113
Add to Cart
Add to Cart
Origami B host strains carry the same trxB/gor mutations as the original Origami strains, except that they are derived from a lacZY mutant of BL21.
Thus the Origami B strains combine the desirable characteristics of BL21, Tuner™ and Origami hosts in one strain background. The trxB and gor mutations are selectable on kanamycin and tetracycline, respectively; therefore, this strain is recommended for use with pET plasmids carrying the ampicillin resistance marker bla.
DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage ≤-70°C
Quantity 1 Set