Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Origami™ B (DE3) pLysS Competent Cells

Catalog No. 708394 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.4 mL
1 mL
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
70-839-4 1 mL
70-839-3 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 70-839-4 Supplier MilliporeSigma™ Supplier No. 708394
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Origami B host strain derived from a lacZY mutant of BL21 to enable precise control of expression levels using IPTG. T7 lysozyme expression suppresses basal T7 expression.

Origami™ B host strains carry the same mutations in trxB and gor as the original Origami strains, except that they are derived from a lacZY mutant of BL21 to enable precise control of expression levels by adjusting the concentration of IPTG. Thus the Origami B strains combine the desirable characteristics of BL21, Tuner™, and Origami strains in one strain background. The mutations in trxB and gor are selectable on kanamycin and tetracycline, respectively; therefore, these strains cannot be used with plasmids that can only selected with kanamycin or tetracycline. These strains also include the lon and ompT deficiences of BL21, which increase protein stability. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG. pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
TRUSTED_SUSTAINABILITY

Specifications

Efficiency Guaranteed efficiency: >2 x 106 cfu/μg
Content And Storage ≤-70°C
Quantity 1 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.