Learn More
MilliporeSigma™ Novagen™ Rosetta-gami™ 2 Competent Cell Set
Catalog No. 714323 Shop All MilliporeSigma Products
Change view
Click to view available options
Content And Storage:
≤-70°C
Catalog No. Content And Storage
71-432-3 ≤-70°C
1 options
Catalog No. 71-432-3 Supplier MilliporeSigma™ Supplier No. 714323
Add to Cart
Add to Cart
Novagen's Rosetta™ 2 host strains are BL21 derivatives designed to enhance the expression of eukaryotic proteins that contain codons rarely used in E. Coli.
Rosetta-gami 2 host strains combine the advantages of Rosetta 2 and Origami 2 strains to alleviate codon bias and enhance disulfide bond formation in the cytoplasm when heterologoous proteins are expressed in E. coli. These trxB/gor mutants are compatible with kanamycin-resistant vectors, and carry the chloramphenicol-resistant pRARE2 plasmid, which supplies seven rare tRNAs. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG. pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
TRUSTED_SUSTAINABILITY
Specifications
Efficiency Guaranteed efficiency: 2 x 106 cfu/μg
Content And Storage ≤-70°C
Quantity 1 Set