Learn More
MilliporeSigma™ Novagen™ Rosetta-gami™ B Competent Cell Set
Catalog No. 711773 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
1 Set
Catalog No. Quantity
71-177-3 1 Set
1 options
Catalog No. 71-177-3 Supplier MilliporeSigma™ Supplier No. 711773
Add to Cart
Add to Cart
Rosetta-gami B strains combine the key features of BL21, Origami, and Rosetta to enhance the expression of eukaryotic proteins. Contains the pLacI plasmid producing extra Lac repressor.
Rosetta-gami B strains combine the key features of BL21 (and its Tuner™ derivative), Origami, and Rosetta to enhance both the expression of eukaryotic proteins and the formation of target protein disulfide bonds in the bacterial cytoplasm. These strains are compatible with ampicillin- or spectinomycin-resistant vectors. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG. pLysS strains express T7 lysozyme, which further suppresses basal expression of T7 RNA polymerase prior to induction, thus stabilizing pET recombinants encoding target proteins that affect cell growth and viability.
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage ≤-70°C
Quantity 1 Set