Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Novagen™ Rosetta-gami™ B (DE3) pLacI Competent Cells

Catalog No. 711383 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
0.4 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-138-3 0.4 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 71-138-3 Supplier MilliporeSigma™ Supplier No. 711383
Add to Cart
Add to Cart

missing translation for 'validMainframeMsg'

Rosetta-gami B strains combine the key features of BL21, Origami, and Rosetta to enhance the expression of eukaryotic proteins. Contains the pLacI plasmid producing extra Lac repressor.

Expression host strains for pETBlue™ and pTriEx™ vectors are lDE3 lysogens that also carry the compatible pLacI plasmid, encoding the lac repressor. The pLacI plasmid confers chloramphenicol resistance and provides enough lac repressor to inhibit transcription from the T7lac promoter in the absence of inducer. The NovaBlue host strain is recommended for initial cloning with pETBlue and pTriEx vectors due to its superior transformation efficiency and high yields of high-quality plasmid DNA. Note that these strains are not recommended for use with pET, pACYCDuet™, pCDF, pRSF, pCOLADuet™ or pETcoco™ vectors. Rosetta-gami B strains combine the key features of BL21 (and its Tuner™ derivative), Origami, and Rosetta to enhance both the expression of eukaryotic proteins and the formation of target protein disulfide bonds in the bacterial cytoplasm. These strains are compatible with ampicillin- or spectinomycin-resistant vectors. In Rosetta-gami B(DE3)pLacI, the rare tRNA genes are present on the same plasmid that carries the lac repressor gene. DE3 indicates that the host is a lysogen of λDE3, and therefore carries a chromosomal copy of the T7 RNA polymerase gene under control of the lacUV5 promoter. Such strains are suitable for production of protein from target genes cloned in pET vectors by induction with IPTG.
TRUSTED_SUSTAINABILITY

Specifications

Content And Storage ≤-70°C
Quantity 0.4 mL
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.