Learn More
MilliporeSigma™ Novagen™ S-protein HRP Conjugate
Catalog No. 690473MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μL
Catalog No. Quantity
69-047-3MI 50 μL
1 options
Catalog No. 69-047-3MI Supplier MilliporeSigma™ Supplier No. 690473
Add to Cart
Add to Cart
S-protein conjugated to HRP
  • Optimized to give highest signal:noise ratio when detecting S·Tag fusion proteins even when present in low abundance in complex protein mixtures
  • Allows single step detection without the need for secondary reagents
  • Highly specific for S·Tag fusion proteins and can be used in the same incubation mix with antibodies or steptavidin conjugates
TRUSTED_SUSTAINABILITY
Specifications
Product Type S-protein HRP Conjugate
Quantity 50 μL
Content And Storage -20°C