Learn More
MilliporeSigma™ Novagen™ T7·Tag™ Antibody HRP Conjugate
Catalog No. 690483MI Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
100 μL
Catalog No. Quantity
69-048-3MI 100 μL
1 options
Catalog No. 69-048-3MI Supplier MilliporeSigma™ Supplier No. 690483
Add to Cart
Add to Cart
Single detection reagent that eliminates need for secondary antibodies or streptavidin conjugates
The T7 Tag Antibody HRP Conjugate is useful as a single detection reagent and eliminates the need for secondary antibodies or streptavidin conjugates.The enzyme conjugated T7 Tag monoclonal antibody enables detection with only one binding step and eliminates cross-reactivity associated with secondary reagents. The T7 Tag Antibody is a mouse monoclonal antibody (IgG2b) directed against the 11 amino acid gene 10 leader peptide expressed by many of the pET translation vectors as well as pSCREEN™ and pRSET vectors. Because the peptide is the natural amino terminal end of the T7 major capsid protein, the antibody also recognizes T7 bacteriophage (e.g., the T7Select™ Vectors).
TRUSTED_SUSTAINABILITY
Specifications
Content And Storage -20°C
Product Type T7·Tag™ Antibody HRP Conjugate