Learn More
MilliporeSigma™ Novagen™ Veggie NovaBlue Singles™ Competent Cells
Catalog No. 712514 Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
11 Reactions
22 Reactions
2 product options available for selection
Product selection table with 2 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
71-251-4 22 Reactions
71-251-3 11 Reactions
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 2 options available.
2 options
Catalog No. 71-251-4 Supplier MilliporeSigma™ Supplier No. 712514
Add to Cart
Add to Cart
Produces high-quality plasmid DNA
Veggie™ NovaBlue Singles™ Competent Cells are available for routine cloning applications. NovaBlue is a K-12 strain ideally suited as an initial cloning host due to its high transformation efficiency, blue/white screening capability (with appropriate plasmids), and mutations in recA and endA, which result in high yields of high-quality plasmid DNA.
TRUSTED_SUSTAINABILITY
Specifications
Efficiency Guaranteed efficiency: >1.5 x 108 cfu/μg
Content And Storage ≤-70°C
Quantity 22 Reactions