Learn More
Description
Specifications
Specifications
| Formulation | Lyophilized from 0.2μM filtered PBS, BSA. |
| Preparation Method | Reconstitute to 100μg/mL with sterile PBS containing ≥1% human or bovine serum albumin. |
| Purification Method | SDS-PAGE |
| Quantity | 25 μg |
| Immunogen | NRG1 Isoform SMDF |
| Storage Requirements | Following reconstituion refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −20°C. |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Recombinant |
| Form | Lyophilized |
| Show More |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.