Learn More
Compatible with a broad range of solvents
  • Hydrophilic
  • Plain surface
  • Sterilization: Gamma or EO compatible
  • Operating Temperature: 75°C max.
  • Color: white
  • Mesh openings range from 10 to 180μm
  • Ideal for collection of algae and cells, particle analysis, large particulate filtration, background filter for automated particle imaging systems, prefiltration of solvents, paint monitoring