Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Oncostatin M, Human, Recombinant Protein, E. coli

Catalog No. 49626010UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-626-010UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 49-626-010UG Supplier MilliporeSigma™ Supplier No. 49626010UG
Recombinant, human oncostatin M expressed in E. coli. Pleiotropic cytokine which inhibits growth of A375 melanoma cells and mouse M1 myeloid leukemic cells. Enhances the growth of both endothelial and hematopoietic cells. Has also been reported to have growth stimulatory activities on normal fibroblasts, AIDS-Kaposis sarcoma cells, and a human erythroleukemic cell line, TF-1.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from sterile-filtered 30% acetonitrile, 0.1% TFA, 50μg BSA/μg rhOSM.
Preparation Method Reconstitute to a final concentration of ≥10μg/mL in sterile PBS containing ≥0.1% BSA or HSA.
Purification Method SDS-PAGE
Quantity 10 μg
Source E. coli
Immunogen Oncostatin M
Storage Requirements Following reconstitution, aliquot and freeze (−70°C). Avoid freeze/thaw cycles of solutions. Stock solutions are stable for up to 3 months at −70°C.
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥97%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.