Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Opticap™ XL2 Disposable Capsule Filters with Polysep II™ Media

Catalog No. KGW6A02HH3 Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size:
1.0 + 0.2 μm
1.0 + 0.5 μm
2.0 + 1.2 μm
Inlet Style:
0.56 in. (14 mm) Hose Barb
0.75 in. (19mm) Sanitary Flange
1.5 in. (38mm) Sanitary Flange
Outlet Style:
0.56 in. (14 mm) Hose Barb
0.75 in. (19mm) Sanitary Flange
1.5 in. (38mm) Sanitary Flange
9 product options available for selection
Product selection table with 9 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Inlet Style Outlet Style
KGW6A02HH3 1.0 + 0.5 μm 0.56 in. (14 mm) Hose Barb 0.56 in. (14 mm) Hose Barb
KGW6A02TT3 1.0 + 0.5 μm 1.5 in. (38mm) Sanitary Flange 1.5 in. (38mm) Sanitary Flange
KGW3A02TT3 1.0 + 0.2 μm 1.5 in. (38mm) Sanitary Flange 1.5 in. (38mm) Sanitary Flange
KGW6A02FF3 1.0 + 0.5 μm 0.75 in. (19mm) Sanitary Flange 0.75 in. (19mm) Sanitary Flange
KGW3A02FF3 1.0 + 0.2 μm 0.75 in. (19mm) Sanitary Flange 0.75 in. (19mm) Sanitary Flange
KGW9A02FF3 2.0 + 1.2 μm 0.75 in. (19mm) Sanitary Flange 0.75 in. (19mm) Sanitary Flange
KGW9A02HH3 2.0 + 1.2 μm 0.56 in. (14 mm) Hose Barb 0.56 in. (14 mm) Hose Barb
KGW9A02TT3 2.0 + 1.2 μm 1.5 in. (38mm) Sanitary Flange 1.5 in. (38mm) Sanitary Flange
KGW3A02HH3 1.0 + 0.2 μm 0.56 in. (14 mm) Hose Barb 0.56 in. (14 mm) Hose Barb
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 9 options available.
9 options
Catalog No. KGW6A02HH3 Supplier MilliporeSigma™ Supplier No. KGW6A02HH3
Add to Cart
Add to Cart

Ideal for a broad range of aqueous-based applications

  • Media consists of a borosilicate glass layer and a layer of mixed esters of cellulose membrane
  • Depth of the microfiber filter layer provides high particle loading capacity, and retention of large particulates while maintaining high flow rates
  • Excellent thermal and hydraulic stress resistance
  • Capsule design with pleated Polysep II media minimizes hold-up volume and reduces production losses

Not in-line steam sterilizable

Laboratory Filtration; Buffer Preparation; Cell Culture Media; Large Volume Parenterals (LVP); Ophthalmics; Serum

Order Info

Meets USP Oxidizable Substances Test requirements after a water flush of ≥1L.

TRUSTED_SUSTAINABILITY

Specifications

Autoclavable Autoclavable
Material (Membrane) MCE
Pore Size 1.0 + 0.5 μm
Sterility Non-sterile
Filtration Area 0.06 m2
For Use With (Application) Laboratory Filtration; Buffer Preparation; Cell Culture Media; Large Volume Parenterals (LVP); Ophthalmics; Serum
Inlet Style 0.56 in. (14 mm) Hose Barb
Outlet Style 0.56 in. (14 mm) Hose Barb
Length (English) 5.6 in.
Length (Metric) 14.2 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.