Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ OptiScale™ Capsule Filters for Aseptic Processing Applications

Catalog No. SHVEA47FH3 Shop All MilliporeSigma Products
Change view
Click to view available options
Pore Size:
0.1 μm
0.2 μm
0.22 μm
0.45/0.22 μm
0.5/0.2 μm
Length (English):
2.9 in.
2.91 in.
3.2 in.
3.24 in.
3.7 in.
Length (Metric):
7.4 cm
8.2 cm
9.4 cm
12 product options available for selection
Product selection table with 12 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Pore Size Length (English) Length (Metric)
SHVEA47FH3 0.1 μm 3.24 in. 8.2 cm
SHGEA47HH3 0.5/0.2 μm 3.7 in. 9.4 cm
SHGLA47FH3 0.45/0.22 μm 3.2 in. 8.2 cm
SHGLA47HH3 0.45/0.22 μm 3.7 in. 9.4 cm
SHGLA47FF3 0.45/0.22 μm 2.91 in. 7.4 cm
SHGEA47FF3 0.2 μm 2.9 in. 7.4 cm
SHGEA47FH3 0.2 μm 3.24 in. 8.2 cm
SHVEA47HH3 0.1 μm 3.7 in. 9.4 cm
SHVEA47FF3 0.1 μm 2.9 in. 7.4 cm
SVGLA47FF3 0.22 μm 2.91 in. 7.4 cm
SVGLA47FH3 0.22 μm 3.2 in. 8.2 cm
SVGLA47HH3 0.22 μm 3.7 in. 9.4 cm
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 12 options available.
12 options
Catalog No. SHVEA47FH3 Supplier MilliporeSigma™ Supplier No. SHVEA47FH3
Add to Cart
Add to Cart

Small-volume option for process screening and scaling

  • With “drop-in” filters
  • Non-sterile
  • PVDF vent cap
  • Wettability: Hydrophilic
  • Housing and drainage disk: Polycarbonate
  • Filter support: Polypropylene
  • Filtration Area: 17.7cm2

Cell culture media; Media additives; Bacterial cell processing; Buffer preparations; Ophthalmic solutions; Pharmaceutical facility water; Vaccines; Process intermediates

TRUSTED_SUSTAINABILITY

Specifications

Pore Size 0.1 μm
Sterility Non-sterile
Filtration Area 17.7 cm2
For Use With (Application) Cell culture media; Media additives; Bacterial cell processing; Buffer preparations; Ophthalmic solutions; Pharmaceutical facility water; Vaccines; Process intermediates
Housing Material Polycarbonate
Length (English) 3.24 in.
Length (Metric) 8.2 cm
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.