Learn More
MilliporeSigma™ OptiScale™ Capsule Filters for Aseptic Processing Applications
Shop All MilliporeSigma ProductsDescription
- With “drop-in” filters
- Non-sterile
- PVDF vent cap
- Wettability: Hydrophilic
- Housing and drainage disk: Polycarbonate
- Filter support: Polypropylene
- Filtration Area: 17.7cm2
Cell culture media; Media additives; Bacterial cell processing; Buffer preparations; Ophthalmic solutions; Pharmaceutical facility water; Vaccines; Process intermediates
Specifications
Specifications
| Pore Size | 0.1 μm |
| Sterility | Non-sterile |
| Filtration Area | 17.7 cm2 |
| For Use With (Application) | Cell culture media; Media additives; Bacterial cell processing; Buffer preparations; Ophthalmic solutions; Pharmaceutical facility water; Vaccines; Process intermediates |
| Housing Material | Polycarbonate |
| Length (English) | 3.7 in. |
| Length (Metric) | 9.4 cm |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.