Learn More
MilliporeSigma™ OptiScale™ Capsule Filters for Clarification and Prefiltration with Polysep II Media
Shop All MilliporeSigma ProductsDescription
- Scalable performance
- Disposable
- Choice of media optimizes your process development
- Peel and stick label for easy tracking
- Faster than 47mm disc method
- With “drop-in” filter devices
- Structural components: Polycarbonate
- Filter Support: Polypropylene
- Vent cap: PVDF
- Seal ring: Fluoroelastomer
- Membrane: Durapore™ hydrophilic PVDF
- Milligard: mixed cellulose esters
- Lifegard: Borosilicate glass fiber
Specifications
Specifications
| Material (Membrane) | PVDF |
| Pore Size | 2.0/1.2 μm |
| Filtration Area | 17.7 cm2 |
| For Use With (Application) | For evaluating media to clarify biological feed streams like cell cultures, serum, plasma and vaccines containing high levels of colloidal particles |
| Inlet Style | 19mm (0.75 in.) sanitary flange |
| Outlet Style | 6mm (0.25 in.) hose barb |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.