Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Osteopontin, HisTag™, Mouse, Recombinant Protein, Mouse

Catalog No. 49926050UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
49-926-050UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 49-926-050UG Supplier MilliporeSigma™ Supplier No. 49926050UG
Add to Cart
Add to Cart
Recombinant, mouse osteopontin fused at the C-terminus to a His·Tag™ sequence and expressed in NSO-derived Leu17-Asn294 (Glu99Gly) murine myeloma cells. A secreted, highly acidic, Ca2+-binding, RGD-containing, phosphorylated glycoprotein. Chemotactic for macrophages, smooth muscle cells, endothelial cells, and glial cells. Has been shown to inhibit nitric oxide production and cytotoxicity by activated macrophages. Plays a significant role in inducing granulomas of various origin. OPN is also involved in the migration of embryonic fibroblasts, macrophages, and metastatic cells.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 0.2μM sterile-filtered PBS, 50μg BSA/μg rmOPN.
Preparation Method Reconstitute to ≥100μg/mL with sterile PBS.
Purification Method SDS-PAGE
Quantity 50 μg
Immunogen Osteopontin
Storage Requirements Following reconstitution, aliquot and freeze (−70°C). Stock solutions are stable for up to 1 month at 4°C and for up to 3 months at −70°C. Avoid freeze/thaw cycles of stock solutions.
Regulatory Status RUO
Conjugate His-Tag
Species Mouse
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.