Learn More
MilliporeSigma™ p34 cdk1/Cyclin B, Human, Recombinant Protein, S. frugiperda
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | In 100mM NaCl, 50mM HEPES, 1mM DTT, 100μM EDTA, 50% glycerol, 0.01% BRIJ™ 35 Detergent, pH 7.5. |
| Quantity | 200 U |
| Source | S. frugiperda |
| Immunogen | p34cdk1/Cyclin B |
| Storage Requirements | Following initial thaw, aliquot and freeze (−70°C). |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Recombinant |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.