Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ p53, HisTag™, Human, Recombinant Protein, E. coli

Catalog No. 50616510UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
50-616-510UG 10 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 50-616-510UG Supplier MilliporeSigma™ Supplier No. 50616510UG
Recombinant, human, wild-type p53 fused at the N-terminus to a His·Tag™ sequence and expressed in E. coli. p53, a tumor suppressor, blocks transformation and inhibits tumor cell growth. Numerous mutations in the p53 gene have been characterized in human cancers. Loss or mutation of p53 function is highly correlated with tumorigenesis. The purified protein is useful in DNA binding assays and as a protein kinase substrate.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 50mM KCl, 30mM sodium pyrophosphate, 5% glycerol, pH 7.5.
Purification Method SDS-PAGE
Quantity 10 μg
Source E. coli
Immunogen p53
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Conjugate His-Tag
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥80%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.