Learn More
MilliporeSigma™ Phosphodiesterase 3A, Active, Human, Recombinant Protein, S. frugiperda
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | In 150mM NaCl, 10mM Tris, 0.1% NP-40, pH 8.3. |
| Quantity | 10 U |
| Source | S. frugiperda |
| Immunogen | Phosphodiesterase 3A |
| Storage Requirements | Following reconstitution, aliquot and freeze (−70°C). |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Recombinant |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.