Learn More
MilliporeSigma™ Phosphodiesterase 5, Catalytic Domain, Human, Recombinant Protein, S. frugiperda
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | In 150mM NaCl, 10mM Tris-HCl, 0.1% NP-40, pH 8.3. |
| Quantity | 10 U |
| Source | S. frugiperda |
| Immunogen | Phosphodiesterase 5, Catalytic Domain |
| Storage Requirements | Following initial thaw, aliquot and freeze (−70°C). |
| Regulatory Status | RUO |
| Species | Human |
| Recombinant | Recombinant |
| Form | Liquid |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.