Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Phosphodiesterase 7A1, Human, Recombinant Protein, S. frugiperda

Catalog No. 52475110U Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
10 U
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-475-110U 10 U
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-475-110U Supplier MilliporeSigma™ Supplier No. 52475110U
Add to Cart
Add to Cart
Full-length, recombinant, human phosphodiesterdase 7A1 (PDE7A1) expressed in S. frugiperda insect cells using a baculovirus expression system. PDE7A1 is required for T-cell activation induced by the costimulation of CD3 and CD28 receptors. Increased PDE7A1 correlates with a decrease in cAMP, increased IL-2, and increased cell proliferation. Supplied as a partially purified preparation of catalytically active enzyme useful for inhibitor and activator assays.
TRUSTED_SUSTAINABILITY

Specifications

Formulation In 150mM NaCl, 10mM Tris, 0.1% NP-40, pH 8.3.
Quantity 10 U
Source S. frugiperda
Immunogen Phosphodiesterase 7A1
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.