Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Phospholipase C-γ-2, HisTag™, Human, Recombinant Protein, S. frugiperda

Catalog No. 52518950UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
52-518-950UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 52-518-950UG Supplier MilliporeSigma™ Supplier No. 52518950UG
Full-length, recombinant, human phospholipase C γ-2 fused at the N-terminus to His·Tag™ and S·Tag™ sequences and expressed in S. frugiperda insect cells. This preparation is qualified for use as a substrate for protein tyrosine kinases in in vitro assays. Intrinsic enzyme activity has not been tested. Production of the second messenger molecules diacylglycerol (DAG) and inositol 1,4,5-trisphosphate (IP3) is mediated by activated phosphatidylinositol-specific phospholipase C enzymes, which are crucial enzymes in transmembrane signaling. PLC-γ-2 is phosphorylated on tyrosine residues following ligand-induced activation of a variety of growth factor receptors and immune system receptors, resulting in increased phospholipase activity. The full-length proteins contains the following domains: C2 domain (aa 1059-1152); PH domain (aa 20-131); PI-PLC X-box catalytic domain (aa 312-456); PI-PLC Y-box catalytic domain (aa 930-1044); two SH2 domains (aa 532-635, aa 646-735); and a SH3 domain (aa 769-829). It has been reported to be phosphorylated at Tyr753 and Tyr759 (Btk, Src, Fyn, Lck), and Tyr1217 and Tyr1245 by Bcr-Abl.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In PBS.
Purification Method SDS-PAGE
Quantity 50 μg
Source S. frugiperda
Immunogen Phospholipase C-γ-2
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Conjugate His-Tag
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.