Learn More
MilliporeSigma™ Phospholipase D, Streptomyces chromofuscus, Protein
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Preparation Method | 10mM Tris-HCl, pH 8.0, 0.05% BSA, and 0.1% (w/v) TRITON™ X-100 (1mg/mL) |
| Quantity | 250 U |
| Source | Streptomyces chromofuscus |
| Immunogen | Phospholipase D |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C) for long-term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for up to 6 months at 4°C or for up to 1 year at −20°C. |
| Regulatory Status | RUO |
| Recombinant | Native |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.