Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ PKCδ, Human, Recombinant Protein, S. frugiperda

Catalog No. 5396725UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
5 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-967-25UG 5 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-967-25UG Supplier MilliporeSigma™ Supplier No. 5396725UG
Add to Cart
Add to Cart
Full-length, recombinant, human PKCδ expressed in S. frugiperda insect cells using a baculovirus expression system. Suitable for studies of PKCδ and its regulation. PKCδ is a Ca2+-independent and phospholipid-dependent enzyme.
TRUSTED_SUSTAINABILITY

Specifications

Concentration Concentration on Label
Formulation In 250mM NaCl, 20mM HEPES, 5mM DTT, 2mM EDTA, 2mM EGTA, 50% glycerol, 0.05% TRITON™ X-100 Detergent, pH 7.5.
Purification Method SDS-PAGE
Quantity 5 μg
Source S. frugiperda
Immunogen PKCδ
Storage Requirements Following initial thaw, aliquot and freeze (−70°C).
Regulatory Status RUO
Species Human
Recombinant Recombinant
Form Liquid
Purity or Quality Grade ≥95%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.