Learn More
MilliporeSigma™ Plasminogen, EACA- and Lysine-Free, Human Plasma, Protein
Shop All MilliporeSigma ProductsDescription
Specifications
Specifications
| Formulation | Lyophilized from 20mM sodium phosphate buffer, 10mg D-mannitol, 10mg NaCl. |
| Preparation Method | H2O |
| Quantity | 50 U |
| Immunogen | Plasminogen |
| Storage Requirements | Following reconstitution, aliquot and freeze (−20°C) for long-term storage or refrigerate (4°C) for short-term storage. Stock solutions are stable for at least 1 week at 4°C or for 1-2 months at −20°C. |
| Regulatory Status | RUO |
| Without Additives | EACA- and Lysine-Free |
| Species | Human |
| Recombinant | Native |
| Form | Lyophilized |
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.