Promotional price valid on web orders only. Your contract pricing may differ. Interested in signing up for a dedicated account number?
Learn More

MilliporeSigma™ Progranulin, CF, Mouse, Recombinant Protein

Catalog No. 53700950UG Shop All MilliporeSigma Products
Change view
Click to view available options
Quantity:
50 μg
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Catalog No. Quantity
53-700-950UG 50 μg
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
Catalog No. 53-700-950UG Supplier MilliporeSigma™ Supplier No. 53700950UG
Recombinant, mouse progranulin, consisting of amino acids 18-589, fused at the C-terminus to a His·Tag™ sequence, and expressed in NS0-derived mouse myeloma cells. Progranulin is an ubiquitously expressed, secreted glycoprotein that is involved in regulation of cell proliferation.
TRUSTED_SUSTAINABILITY

Specifications

Formulation Lyophilized from 0.2μM filtered PBS.
Preparation Method Reconstitute to 250μg/mL with sterile PBS.
Purification Method SDS-PAGE
Quantity 50 μg
Immunogen Progranulin
Storage Requirements Following reconstitution refrigerate (4°C) for short-term storage or aliquot and freeze (−20°C) for long-term storage. Stock solutions are stable for up to 1 month at 4°C or for up to 3 months at −20°C.
Regulatory Status RUO
Without Additives Carrier-Free
Species Mouse
Recombinant Recombinant
Form Lyophilized
Purity or Quality Grade ≥90%
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.